missing translation for 'onlineSavingsMsg'
Learn More

Menaquinone-4, MedChemExpress

Product Code. 16489885
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489885 250 mg 250mg
16438856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489885 Supplier Medchem Express Supplier No. HYB2156
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Menaquinone-4 is a vitamin K, used as a hemostatic agent, and also a adjunctive therapy for the pain of osteoporosis.

Menaquinone-4 is a vitamin K, used as a hemostatic agent, and also a adjunctive therapy for the pain of osteoporosis.

Specifications

Chemical Name or Material Menaquinone-4
CAS 863-61-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H40O2
Quantity 250 mg
Synonym Vitamin K2(MK-4) Menaquinone K4
Solubility Information DMSO : 20 mg/mL (44.98 mM; Need ultrasonic)
SMILES O=C1C(C)=C(C/C=C(C)/CC/C=C(C)/CC/C=C(C)/CC/C=C(C)\C)C(C2=C1C=CC=C2)=O
Molecular Weight (g/mol) 444.65
Formula Weight 444.65
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.