missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Menadione bisulfite sodium, MedChemExpress
Description
Menadione bisulfite (sodium) is used as an agent to induce acute oxidative stress, and to function as a plant-defense activator against several pathogens.
Specifications
Specifications
| Chemical Name or Material | Menadione bisulfite sodium |
| CAS | 130-37-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C11H9NaO5S |
| Quantity | 1 g |
| Synonym | Menadione sodium bisulfite Vitamin K3 sodium bisulfite |
| Solubility Information | H2O : ≥ 100 mg/mL (362.00 mM) ∣DMSO : 100 mg/mL (362.00 mM; Need ultrasonic) |
| SMILES | O=S(C(CC(C1=C2C=CC=C1)=O)(C)C2=O)(O[Na])=O |
| Molecular Weight (g/mol) | 276.24 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?