missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Membrin Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | Membrin |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18321957
|
Novus Biologicals
NBP3-03314-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18337486
|
Novus Biologicals
NBP3-03314-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Membrin Polyclonal specifically detects Membrin in Human samples. It is validated for Western Blot.Specifications
| Membrin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| golgi SNAP receptor complex member 2, GS27Bos1,27 kDa Golgi SNARE protein, membrin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human Membrin (NP_004278.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Membrane Trafficking and Chaperones | |
| PBS with 50% glycerol, pH7.3. | |
| 9570 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title