missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MembraneMax™ Protein Expression Kit
Description
MembraneMax™ Protein Expression Kits delivers high yields of soluble (dispersed) membrane proteins using the MembraneMax™ reagent, which is an planar phospholipid membrane bilayer surrounded by a scaffold protein (also called a nanolipoprotein particle or NLP). Combining this technology with a scalable cell-free expression E. coli extract delivers microgram to milligram quantities of your membrane protein specifically for high throughput expression screening, expression of membrane proteins that would otherwise be toxic in cell-based systems and functional or activity assays. With high uniformity and consistent structure, MembraneMax reagent minimizes formation of membrane protein aggregates or clumping often caused by inconsistent size, shape and structure typical observed with microsomes or micelles. The MembraneMax™ Protein Expression kits contain all the necessary reagents for successful expression of your membrane protein.
Key Advantages of the MembraneMax™ Protein Expression Kits:
Optimized cell-free expression delivers high yield of membrane protein from nanogram to milligram quantities
MembraneMax™ reagent enables a monodispersed population of soluble membrane protein-NLP complexes
Expression format amenable to high-throughput protein synthesis for screening and expression of toxic proteins
Production of a homogeneous population of your membrane protein
Convenient kit format includes the necessary reagents for protein expression—simply add your gene of interest to get started
Key Advantages of the MembraneMax™ Protein Expression Kits:
Specifications
Specifications
| Description | MembraneMax™ Protein Expression Kit, 20 rxns, T7 Promoter, Membrane Protein Expressed Protein Type, Not High Throughput-Compatible, Untagged Protein Tag, Used For Protein Expression |
| For Use With (Application) | Protein expression |
| Includes | Expressway™ mini expression module, Expressway™ mini amino acids module, MembraneMax™ expression module, Expressway™ (extract) module, Expressway™ amino acids module and MembraneMax™, expression modules |
| Sample Volume | 20 rxn(s) |
| Product Type | MembraneMax™ Protein Expression Kit |
| Quantity | 20 rxn(s) |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?