Learn More
Carl Zeiss™ Membrane Slides
Product Code. 15350731
Change view
Click to view available options
Coating:
PEN Membrane
Quantity:
100 Pack
Unit Size:
Pack of 100
Product Code. Coating Quantity Unit Size
15350731 PEN Membrane 100 Pack Pack of 100
1 options
Product Code. 15350731 Supplier Carl Zeiss™ Supplier No. 4151909041000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Membrane-covered microscope slides come ready to use with a white marking area.
  • Membrane-covered
  • Ready to use
  • White marking area
Specifications
Type Membrane Slide
Coating PEN Membrane
Description 1.0 PEN (D) PEN membrane-covered
Quantity 100 Pack