missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM (Temin's modification) (2X), no phenol red
GREENER_CHOICE

Product Code. 12037549
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12037549 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 12037549 Supplier Gibco™ Supplier No. 21935028
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

MEM (Temin's modification) (2X), no phenol red

Temin's Modified Eagle Medium, (MEM) is a modified form of Minimum Essential Medium, one of the most commonly used of all cell culture media. MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including Glasgow's MEM, MEM α, DMEM, and Temin's Modification. Temin's Modification is a 2x MEM with 4x the vitamins and amino acids of the original MEM. This MEM formulation contains Earle's salts for use in a CO2 incubator.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ MEM is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

MEM contains no proteins, lipids, or growth factors. Therefore, MEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (8.4 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 X
Form Liquid
Product Type MEM (Temin's Modification) Media
With Additives Sodium Bicarbonate
Without Additives No HEPES, No Phenol Red
For Use With (Application) Mammalian cell culture
Quantity 250 mL
Certifications/Compliance ISO 13485 standards, FDA
Regulatory Status In-vitro Diagnostic Use
Content And Storage 2 to 8°C
Description MEM (Temin's modification) Media, 2X Concentration, 250mL, Liquid Form, With L-glutamine, Sodium Bicarbonate, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Soluble in Water, Used for Mammalian Cell Culture
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.