missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ MEM Rega-3

Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
Minimum Essential Medium (MEM) REGA-3 is a modification of the commonly used MEM. The Rega-3 modification is often used to culture HEP-G2 (human liver carcinoma) cells and occasionally other human tumor cells. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including this Rega-3 modification. MEM Rega-3 is unique because it has a reduced sodium bicarbonate concentration, modified salt concentrations, and contains the antibiotic gentamycin sulfate. This product is made with Earle’s salts.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
cGMP Manufacturing and Quality System
Gibco™ MEM is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
MEM Rega-3 contains no proteins, lipids, or growth factors. Therefore, MEM Rega-3 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (0.85 g ⁄ L) and therefore requires a low CO2 environment to maintain physiological pH.
Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including this Rega-3 modification. MEM Rega-3 is unique because it has a reduced sodium bicarbonate concentration, modified salt concentrations, and contains the antibiotic gentamycin sulfate. This product is made with Earle’s salts.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
cGMP Manufacturing and Quality System
Gibco™ MEM is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
MEM Rega-3 contains no proteins, lipids, or growth factors. Therefore, MEM Rega-3 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (0.85 g ⁄ L) and therefore requires a low CO2 environment to maintain physiological pH.
Specifications
Specifications
| Form | Liquid |
| Product Type | MEM Rega-3 |
| With Additives | Phenol Red, Gentamicin, Sodium Bicarbonate |
| Without Additives | No Glutamine, No HEPES |
| For Use With (Application) | Mammalian cell culture |
| Quantity | 500 mL |
| Certifications/Compliance | ISO 13485 standards, FDA |
| Regulatory Status | For research use only |
| Content And Storage | 2 to 8°C, room temperature |
| Description | MEM Rega-3, 500mL, Liquid, 12 Months Shelf Life, Gentamicin Included Antibiotics, Ambient Shipping Condition, 2 to 8°C Storage Requirements, Fewer Resources Used, Less waste Green Features, For Mammalian Cell Culture |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction