missing translation for 'onlineSavingsMsg'
Learn More

Sartorius MEM Eagle for Suspension

Product Code. 31908436 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908436 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908436 Supplier Sartorius Supplier No. 010451A
 Restricted Item
Estimated Shipment: 23-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MEM is used for the cultivation of a wide array of cells grown in monolayers.

  • Minimum Essential Medium (MEM) is one of the most commonly used cell culture media.
  • MEM can be used with a variety of suspension and adherent mammalian cells, including HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, fibroblasts, and primary rat astrocytes.
  • Modified as follows:
    • With Glucose
    • With Magnesium
    • With Earle's Salts
    • With Sodium Bicarbonate
    • With Phenol Red
    • Without L-Glutamine

Tekniske data

Classification Chemically-defined, Animal Origin-free
Form Liquid
Product Type MEM Cell Culture Media
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Glucose, Magnesium, Earle's Balanced Salt Solution (EBSS), Sodium Bicarbonate, Phenol Red
Without Additives No Glutamine
Quantity 500 mL
Culture Type Monolayer Culture
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.