missing translation for 'onlineSavingsMsg'
Learn More

Sartorius MEM-Eagle with Non-Essential Amino Acids, without L-Glutamine

Product Code. 31908369 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908369 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908369 Supplier Sartorius Supplier No. 010401A
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MEM is used for the cultivation of a wide array of cells grown in monolayers.

  • MEM-Eagle with Non-Essential Amino Acids (MEM-NEAA), without L-Glutamine, is one of the most widely utilized synthetic growth media.
  • Optional supplementation with Non-Essential Amino Acids (NEAAs) in formulations containing Hank's or Eagle's salts broadens the utility of this medium.
  • MEM is a modification of BME containing higher concentrations of essential nutrients.
  • MEM supports the cultivation of a broad range of monolayer cell cultures.
  • Research by Eagle and others demonstrated that additions to the original BME formulation could support a wider spectrum of fastidious cells in culture.
  • MEM incorporates these modifications and includes higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells in culture.
  • MEM-NEAA supplementation provides nutrients that may reduce the overall biosynthetic burden on cell cultures in vitro.
  • Modified as follows:
    • With Earle's Salts
    • With Non-essential Amino Acids
    • With Phenol Red
    • Without L-Glutamine

Spezifikation

Classification Chemically-defined, Animal Origin-free
Form Liquid
Product Type MEM Cell Culture Media
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Earle's Balanced Salt Solution (EBSS), Non Essential Amino Acids (NEAA), Phenol Red
Without Additives No Glutamine
Quantity 500 mL
Culture Type Monolayer Culture
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.