Learn More
Stuart™ Melting Point Standard Kit
Shop All Cole-Parmer Products
Click to view available options
Display Type:
Large 7 in. Touchscreen Display
Description
- Capture and display video image of the melt on the touch-screen display or download to a PC.
- Accurately identify the melting point with advanced digital image processing.
- Get up and running quickly - intuitive setup and navigation on large 7 in. color touch screen.
- Increase productivity - analyse three samples simultaneously.
- Take measurements, view results, save programs, set up user sign-ins, and more.
- Rapid ramp mode to take a fast scan of the full temperature range.
- Watch the melt on the display, or set the unit up and walk away and view a video of the melt later.
- Download videos and reports to a USB flash drive or use the optional printer for direct printing of results.
- Stored video results include a temperature stamp of the captured melt temperature.
- Storage compartment for unused capillaries includes a glass cutter to open capillaries that are closed on both ends.
- Hold-area will keep three prepared sample capillaries safely away from the heat source.
- Conforms to US Pharmacopeia (USP) and GLP requirements.
Specifications
Specifications
| Product Type | Advanced Melting Point Apparatus |
| Certifications/Compliance | Conforms to US Pharmacopeia (USP) and GLP requirements |
| Display Type | Large 7 in. Touchscreen Display |
| Includes | Glass Capillaries, Power Cord, Glass Cutter |