missing translation for 'onlineSavingsMsg'
Learn More

Melphalan, Tocris Bioscience™

Product Code. 30001644
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30001644

Brand: Tocris Bioscience 4619/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

DNA alkylating agent; cytotoxic and antineoplastic

DNA alkylating agent; induces cytotoxicity through the formation of stable interstrand and intrastrand crosslinks within DNA. Inhibits growth of PC-3 cells (IC50 values are 0.074 and 0.77 μg/ml for sequential dosing and single dosing respectively).
TRUSTED_SUSTAINABILITY

Specifications

CAS 148-82-3
Molecular Formula C13H18Cl2N2O2
Quantity 50 mg
Synonym melphalan, alkeran, melfalan, phenylalanine mustard, l-sarcolysine, l-pam, l-sarcolysin, l-phenylalanine mustard, levofalan, p-l-sarcolysin
InChI Key SGDBTWWWUNNDEQ-LBPRGKRZSA-N
SMILES C1=CC(=CC=C1CC(C(=O)O)N)N(CCCl)CCCl
IUPAC Name (2S)-2-amino-3-[4-[bis(2-chloroethyl)amino]phenyl]propanoic acid
Molecular Weight (g/mol) 305.199
PubChem CID 460612
ChEBI CHEBI:28876
Formula Weight 305.2
Percent Purity >95%
Chemical Name or Material Melphalan
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.