missing translation for 'onlineSavingsMsg'
Learn More

Melphalan, 94%

Product Code. 15572343 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15572343

Brand: Thermo Scientific Acros 458650050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Melphalan, CAS # 148-82-3, belongs to the class of nitrogen mustards with anti-neoplastic activity. | CAS: 148-82-3 | C13H18Cl2N2O2 | 305.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Melphalan is a phenylalanine derivative of nitrogen mustard with antineoplastic activity with the capacity to alkylate nucleic acids
  • Melphalan alkylates DNA at the N-7 position of guanine and induces DNA interstrand cross-linkages, resulting in the inhibition of DNA/RNA synthesis, inducing cytotoxicity against both dividing and non-dividing tumor cells

Applications

  • Melphalan has been used in in vitro experiments to treat multiple myeloma in cell lines and breast cancer cell lines
  • In in vivo experiments, using rats, it shows a potent therapeutic effect in cases of sarcomatous pulmonary metastases
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148-82-3
Molecular Formula C13H18Cl2N2O2
Molecular Weight (g/mol) 305.2
InChI Key SGDBTWWWUNNDEQ-LBPRGKRZSA-N
Synonym melphalan, alkeran, melfalan, phenylalanine mustard, l-sarcolysine, l-pam, l-sarcolysin, l-phenylalanine mustard, levofalan, p-l-sarcolysin
PubChem CID 460612
ChEBI CHEBI:28876
IUPAC Name (2S)-2-amino-3-[4-[bis(2-chloroethyl)amino]phenyl]propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)N(CCCl)CCCl

Specifications

Melting Point 180.0°C
Infrared Spectrum Authentic
Assay Percent Range 93% min. (HPLC)
Linear Formula (ClCH2CH2)2NC6H4CH2CH(NH2)COOH
Quantity 5 g
Formula Weight 305.2
Percent Purity 94%
Chemical Name or Material Melphalan
compliance-icons compliance-icons
Product Identifier
  • Melphalan
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H350-May cause cancer.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361-Suspected of damaging fertility or the unborn child.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.