missing translation for 'onlineSavingsMsg'
Learn More

Melperone, MedChemExpress

Product Code. 16488856
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488856 100 mg 100mg
16418748 10 mg 10mg
16404657 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488856 Supplier Medchem Express Supplier No. HYB2169
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Melperone, a butyrophenone, is an antipsychotic drug used for sleep induction which is frequently prescribed in psychiatric setting. Melperone has been used for a variety of indications, including the treatment of schizophrenia, but also for agitation in the elderly.

Melperone, a butyrophenone, is an antipsychotic drug used for sleep induction which is frequently prescribed in psychiatric setting. Melperone has been used for a variety of indications, including the treatment of schizophrenia, but also for agitation in the elderly.

Specifications

Chemical Name or Material Melperone
CAS 3575-80-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H22FNO
Quantity 100 mg
Solubility Information DMSO : 41.67 mg/mL (158.23 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(F)C=C1)CCCN2CCC(C)CC2
Molecular Weight (g/mol) 263.35
Formula Weight 263.35
Percent Purity 95.32%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.