missing translation for 'onlineSavingsMsg'
Learn More

Meloxicam, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J60635.06

Quantity 5g

 View more versions of this product

Product Code. 15473369

  • £57.30 / 5g
 Restricted Item
Estimated Shipment: 13-10-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water (22 mg/ml), DMSO (25 mg/ml), DMF, acetone(slightly soluble), ethanol(slightly soluble), and methanol(slightly soluble).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

71125-38-7
351.40
DWMREKMVXIFPFM-ACCUITESSA-N
54677470
(3E)-3-{hydroxy[(5-methyl-1,3-thiazol-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λ⁶,2-benzothiazine-1,1,4-trione
C14H13N3O4S2
MFCD00868752
meloxicam, mobic, metacam, movalis, meloxicamum, mobicox, mobec, meloxicam mobic, movatec, parocin
CHEBI:6741
CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O

Specifications

71125-38-7
MFCD00868752
UN2811
meloxicam, mobic, metacam, movalis, meloxicamum, mobicox, mobec, meloxicam mobic, movatec, parocin
DWMREKMVXIFPFM-ACCUITESSA-N
(3E)-3-{hydroxy[(5-methyl-1,3-thiazol-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λ⁶,2-benzothiazine-1,1,4-trione
54677470
351.39
C14H13N3O4S2
5g
14,5826
Soluble in water (22mg/ml),DMSO (25mg/ml),DMF,acetone(slightly soluble),ethanol(slightly soluble),and methanol(slightly soluble).
CN1\C(=C(\O)NC2=NC=C(C)S2)C(=O)C2=CC=CC=C2S1(=O)=O
351.40
CHEBI:6741
Meloxicam
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.