missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MELK Antibody (PSH03-55), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32587
This item is not returnable.
View return policy
Description
MELK Monoclonal antibody specifically detects MELK in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| MELK | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| EC 2.7.11, EC 2.7.11.1, hMELK, HPK38, KIAA0175pEg3 kinase, maternal embryonic leucine zipper kinase, Protein kinase PK38 | |
| Recombinant protein within human MELK aa 1-400/ 651. (Uniprot: Q14680) | |
| 100 μL | |
| Protein Kinase, Stem Cell Markers | |
| 9833 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| PSH03-55 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction