missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MELK Antibody (2G2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £435.00
Specifications
| Antigen | MELK |
|---|---|
| Clone | 2G2 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18104538
|
Novus Biologicals
NBP2-37481 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650516
|
Novus Biologicals
NBP2-37481-0.025ml |
0.025 mL |
£173.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MELK Monoclonal specifically detects MELK in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| MELK | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 2.7.11, EC 2.7.11.1, hMELK, HPK38, KIAA0175pEg3 kinase, maternal embryonic leucine zipper kinase, Protein kinase PK38 | |
| MELK | |
| IgG2a | |
| Affinity Purified |
| 2G2 | |
| Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Embryonic Stem Cell Markers, Protein Kinase, Stem Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| 9833 | |
| Synthesized peptide of human MELK(AA: 637-651:C-VYKRLVEDILSSCKV). | |
| Primary | |
| 74.6 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title