missing translation for 'onlineSavingsMsg'
Learn More

MELK-8a hydrochloride, MedChemExpress

Product Code. 16452968
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452968 10 mg 10mg
16475707 5 mg 5mg
16469536 50 mg 50mg
16454286 100 mg 100mg
16430639 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16452968 Supplier Medchem Express Supplier No. HY100368A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MELK-8a hydrochloride is a novel maternal embryonic leucine zipper kinase (MELK) inhibitor with an IC50 of 2 nM.

MELK-8a hydrochloride is a novel maternal embryonic leucine zipper kinase (MELK) inhibitor with an IC50 of 2 nM.

Specifications

Chemical Name or Material MELK-8a hydrochloride
Physical Form Solid
CAS 2096992-20-8
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C25H33ClN6O
For Use With (Application) Cancer-Kinase/protease
Synonym NVS-MELK8a hydrochloride
Solubility Information H2O : ≥ 100 mg/mL (213.21 mM) ∣DMSO : 8.6 mg/mL (18.34 mM; Need ultrasonic and warming)
SMILES CN(CC1)CCN1C2=CC=C(N3C=C(C4=CC=NC=C4OCC5CCNCC5)C=N3)C=C2.Cl
Molecular Weight (g/mol) 469.02
Formula Weight 469.02
Percent Purity 99.26%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.