missing translation for 'onlineSavingsMsg'
Learn More

Melittoside, MedChemExpress

Product Code. 16499188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499188 10 mg 10mg
16491598 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499188 Supplier Medchem Express Supplier No. HYN0915
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Melittoside is a natural compound.

Melittoside is a natural compound.

Specifications

Chemical Name or Material Melittoside
CAS 19467-03-9
Color White
Purity Grade Notes Research
Molecular Formula C21H32O15
Quantity 10 mg
SMILES O[C@H]1[C@@]([C@](C(CO)=C1)([H])[C@@H]2O[C@]([C@@H]([C@@H](O)[C@@H]3O)O)([H])O[C@@H]3CO)(C=CO2)O[C@]([C@@H]([C@@H](O)[C@@H]4O)O)([H])O[C@@H]4CO
Molecular Weight (g/mol) 524.47
Formula Weight 524.47
Percent Purity 98.87%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.