missing translation for 'onlineSavingsMsg'
Learn More

Melatonin, TRC

Product Code. 18030190
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18030190 1 g 1g
18023500 50 mg 50mg
18051710 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18030190 Supplier Toronto Research Chemicals Supplier No. TRCM2150001G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-31-4
Molecular Formula C13 H16 N2 O2
Molecular Weight (g/mol) 232.28
Synonym Melatonin, Acetamide, N-[2-(5-methoxy-1H-indol-3-yl)ethyl]- (9CI, ACI), Acetamide, N-[2-(5-methoxyindol-3-yl)ethyl]- (6CI, 8CI), N-[2-(5-Methoxy-1H-indol-3-yl)ethyl]acetamide (ACI), 3-(2-Acetamidoethyl)-5-methoxyindole, 5-Methoxy-N-acetyltryptamine, Circadin, Melatol, Melatonine, Melaxen, Melovine, N-[2-(5-Methoxyindol-3-yl)ethyl]acetamide, N-Acetyl-5-methoxytryptamine, NSC 113928, NSC 56423, Regulin, Slenyto
IUPAC Name N-[2-(5-methoxy-1H-indol-3-yl)ethyl]acetamide
SMILES COc1ccc2[nH]cc(CCNC(=O)C)c2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H16N2O2/c1-9(16)14-6-5-10-8-15-13-4-3-11(17-2)7-12(10)13/h3-4,7-8,15H,5-6H2,1-2H3,(H,14,16)
Formula Weight 232.1212
Percent Purity >95%
Chemical Name or Material Melatonin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.