missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Melatonin
GREENER_CHOICE

Product Code. 18792514
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792514 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18792514 Supplier Tocris Bioscience™ Supplier No. 3550/50
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Endogenous hormone; agonist at MT1 and MT2

Endogenous hormone that acts as an agonist at melatonin receptors MT1 and MT2. Exhibits a prominent role in the control of circadian rhythm, displays immunomodulatory activity and acts as a powerful antioxidant in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 73-31-4
Quantity 50 mg
Molecular Formula C13H16N2O2
Formula Weight 232.28
Purity >99%
Molecular Weight (g/mol) 232.283
Chemical Name or Material Melatonin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.