missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melatonin Receptor 1B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£321.00 - £526.00
Specifications
| Antigen | Melatonin Receptor 1B |
|---|---|
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18398876
|
Novus Biologicals
NBP3-17202-25UL |
25 μg |
£321.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18384344
|
Novus Biologicals
NBP3-17202-100UL |
100 μg |
£526.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Melatonin Receptor 1B Polyclonal antibody specifically detects Melatonin Receptor 1B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Melatonin Receptor 1B | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuroscience | |
| PBS, pH 7.2, 40% glycerol | |
| 4544 | |
| IgG | |
| Affinity purified |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FGQTL2, Mel 1b melatonin receptor, Mel1b receptor, MEL-1B-R, melatonin receptor 1B, melatonin receptor MEL1B, melatonin receptor type 1B, MT2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: IQDASKGSHAEGLQSPAPPIIGVQHQADAL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title