missing translation for 'onlineSavingsMsg'
Learn More

Melatonin Receptor 1B Antibody, FITC, Novus Biologicals™

Product Code. 18350455 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350455 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350455 Supplier Novus Biologicals Supplier No. NLS932F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Melatonin Receptor 1B Polyclonal specifically detects Melatonin Receptor 1B in Human, Mouse, Rat, Chicken samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Melatonin Receptor 1B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen, Knockdown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P49286
Gene Alias FGQTL2, Mel 1b melatonin receptor, Mel1b receptor, MEL-1B-R, melatonin receptor 1B, melatonin receptor MEL1B, melatonin receptor type 1B, MT2
Gene Symbols MTNR1B
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human Melatonin Receptor 1B protein (within residues 200-300) [Swiss-Prot P49286].
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4544
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.