missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Melanotan II

Product Code. 18735187
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18735187 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18735187 Supplier R&D Systems™ Supplier No. 2566/1
  • ONLINE EXCLUSIVE PRICE
    £167.00
    £157.50 / 1mg
    Save £9.50
    5% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. High affinity melanocortin receptor agonist

Melanotan II is a high affinity melanocortin receptor agonist (Ki values are 0.67, 6.6, 34 and 46 nM for MC1, MC4, MC3 and MC5 receptors respectively). Stimulates erectile activity, inhibits food intake and displays neuroprotective properties in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Product Line Tocris
Purity 0.95
Content And Storage Store at -20°C
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.