missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanopsin Antibody (628737), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10497PEATT594
This item is not returnable.
View return policy
Description
Melanopsin Monoclonal antibody specifically detects Melanopsin in Human samples. It is validated for Flow Cytometry
Specifications
| Melanopsin | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 628737 | |
| Flow Cytometry | |
| melanopsin, MGC142118, MOP, opsin 4 | |
| NS0 mouse myeloma cell line transfected with human Melanopsin, Accession # Q9UHM6 | |
| 0.1 mL | |
| GPCR, Vision | |
| 94233 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction