missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanopsin Antibody (628737) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10497JF585
This item is not returnable.
View return policy
Description
Melanopsin Monoclonal antibody specifically detects Melanopsin in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| Melanopsin | |
| Monoclonal | |
| Janelia Fluor 585 | |
| melanopsin, MGC142118, MOP, opsin 4 | |
| NS0 mouse myeloma cell line transfected with human Melanopsin, Accession # Q9UHM6 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot, CyTOF | |
| 628737 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR, Vision | |
| 94233 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction