missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanophilin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10338-1-AP-20UL
This item is not returnable.
View return policy
Description
Rab effector protein involved in melanosome transport. Serves as link between melanosome-bound RAB27A and the motor protein MYO5A.Specifications
| Melanophilin | |
| Polyclonal | |
| Unconjugated | |
| MLPH | |
| Exophilin 3, melanophilin, MLPH, SLAC2 A, SLAC2A, Synaptotagmin like protein 2a | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79083 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BV36 | |
| MLPH | |
| Melanophilin Fusion Protein Ag0316 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction