missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanoma Marker (MART-1 + gp100) Antibody (DT101+ BC199 + HMB45), PE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Melanoma Marker (MART-1 + gp100) Monoclonal antibody specifically detects Melanoma Marker (MART-1 + gp100) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Specifications
Specifications
| Antigen | Melanoma Marker (MART-1 + gp100) |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Classification | Monoclonal |
| Clone | DT101+ BC199 + HMB45 |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | MART1, MART-1 |
| Host Species | Mouse |
| Immunogen | Recombinant Melanoma Marker (MART-1 + gp100) protein (DT101 & BC199), Extract of pigmented melanoma metastases from lymph nodes (HMB45) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?