missing translation for 'onlineSavingsMsg'
Learn More

Melanoma Marker (MART-1 + gp100) Antibody (DT101+ BC199 + HMB45), Novus Biologicals™

Product Code. 18446101 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18446101 20 μg 20µg
18427211 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18446101 Supplier Novus Biologicals Supplier No. NBP23434120ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Melanoma Marker (MART-1 + gp100) Monoclonal antibody specifically detects Melanoma Marker (MART-1 + gp100) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Melanoma Marker (MART-1 + gp100)
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DT101+ BC199 + HMB45
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 ug/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. Q16655, Q16655, P40967, Q16655
Gene Alias MART1, MART-1
Host Species Mouse
Immunogen Recombinant Melanoma Marker (MART-1 + gp100) protein (DT101 & BC199), Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2315
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG2b κ, IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.