missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanoma Marker (MART-1 + gp100) Antibody (DT101+ BC199 + HMB45), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34608MFV500
This item is not returnable.
View return policy
Description
Melanoma Marker (MART-1 + gp100) Monoclonal antibody specifically detects Melanoma Marker (MART-1 + gp100) in Human samples. It is validated for Flow Cytometry
Specifications
| Melanoma Marker (MART-1 + gp100) | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2315 | |
| Store at 4C in the dark. | |
| IgG2b κ, IgG1 κ |
| Flow Cytometry | |
| DT101+ BC199 + HMB45 | |
| Flow Cytometry | |
| MART1, MART-1 | |
| Recombinant Melanoma Marker (MART-1 + gp100) protein (DT101 & BC199); Extract of pigmented melanoma metastases from lymph nodes (HMB45) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction