missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanoma Associated Antigen Mouse anti-Human, Clone: MAA/1414, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Melanoma Associated Antigen |
|---|---|
| Clone | MAA/1414 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA 1-5 μg/mL |
| Applications | ELISA |
Description
Melanoma Associated Antigen Monoclonal specifically detects Melanoma Associated Antigen in Human samples. It is validated for ELISA.Specifications
| Melanoma Associated Antigen | |
| 0.2 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| proteins extracted from a melanoma | |
| Primary | |
| Store at 4°C. |
| MAA/1414 | |
| ELISA 1-5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| IgG2b κ | |
| Protein A or G purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel