missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanoma Associated Antigen (KBA.62) Antibody (KBA.62), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11535APC
This item is not returnable.
View return policy
Description
Melanoma Associated Antigen (KBA.62) Monoclonal antibody specifically detects Melanoma Associated Antigen (KBA.62) in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
| Melanoma Associated Antigen (KBA.62) | |
| Monoclonal | |
| APC | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| KBA.62 | |
| PBS | |
| Human KAL cells derived from lymph node metastasis of malignant melanoma | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction