missing translation for 'onlineSavingsMsg'
Learn More

Melanoma Associated Antigen (KBA.62) Antibody (IHC062), Novus Biologicals™

Mouse Monoclonal Antibody

£143.00 - £454.00

Specifications

Antigen Melanoma Associated Antigen (KBA.62)
Clone IHC062
Dilution Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18664491
View Documents
Novus Biologicals
NBP2-75984-0.1ml
0.1 mL
£143.00
0.01mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18686420
View Documents
Novus Biologicals
NBP2-75984-1ml
1 mL
£454.00
1mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Melanoma Associated Antigen (KBA.62) Monoclonal antibody specifically detects Melanoma Associated Antigen (KBA.62) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Melanoma Associated Antigen (KBA.62)
Immunohistochemistry 1:100 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200
Monoclonal
Purified
RUO
Tris Buffer, pH 7.3 - 7.7, with 1% BSA
IgG1
Protein A purified
IHC062
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
Human KAL Cells derived from Lymphs
Primary
Store at 4°C in the dark. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.