missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanoma antigen family C2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-68961
This item is not returnable.
View return policy
Description
Melanoma antigen family C2 Polyclonal specifically detects Melanoma antigen family C2 in Human samples. It is validated for Western Blot.
Specifications
| Melanoma antigen family C2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Cancer/testis antigen 10, CT10MAGE-E1 antigen, HCA587MAGE-C2, hepatocellular cancer antigen 587, Hepatocellular carcinoma-associated antigen 587, MAGE-C2 antigen, MAGEE1MGC13377, melanoma antigen family C, 2, melanoma antigen, family E, 1, cancer/testis specific, melanoma-associated antigen C2 | |
| Rabbit | |
| 41 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%. | |
| Human | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9UBF1 | |
| MAGEC2 | |
| Synthetic peptides corresponding to MAGEC2 (melanoma antigen family C, 2) The peptide sequence was selected from the N terminal of MAGEC2. Peptide sequence SCCSSFSWSSFSEESSSQKGEDTGTCQGLPDSESSFTYTLDEKVAELVEF. | |
| Affinity purified | |
| RUO | |
| 51438 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction