missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanoma Antigen Family B16 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Melanoma Antigen Family B16 Polyclonal antibody specifically detects Melanoma Antigen Family B16 in Human samples. It is validated for Immunofluorescence
Specifications
Specifications
| Antigen | Melanoma Antigen Family B16 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
| Formulation | PBS, pH 7.2, 40% glycerol |
| Gene Alias | MAGEB16, MAGE-B16 Antigen, Melanoma Antigen Family B, 16, Melanoma Antigen Family B, 16 (Pseudogene), Melanoma-Associated Antigen B16 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: SEDTSDPRNVPADALDQKVAFLVNFMLHKCQMK |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?