missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanocortin-5 R/MC5R Antibody (821012) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8205JF585
This item is not returnable.
View return policy
Description
Melanocortin-5 R/MC5R Monoclonal antibody specifically detects Melanocortin-5 R/MC5R in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Melanocortin-5 R/MC5R | |
| Monoclonal | |
| Janelia Fluor 585 | |
| MC2, MC-2, MC5-R, melanocortin 5 receptor, melanocortin receptor 5, melanocortin-5 receptor | |
| NS0 mouse myeloma cell line transfected with human Melanocortin-5 R/MC5R, Met1-Asp325, Accession # P33032 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 821012 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 4161 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction