missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanocortin-1 R/MC1R Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£153.00 - £366.00
Specifications
| Antigen | Melanocortin-1 R/MC1R |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18056614
|
Novus Biologicals
NBP3-02970-20UL |
20 μL |
£153.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18360517
|
Novus Biologicals
NBP3-02970-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Melanocortin-1 R/MC1R Polyclonal specifically detects Melanocortin-1 R/MC1R in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| Melanocortin-1 R/MC1R | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CMM5, MC1-R, melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor), Melanocortin receptor 1, melanocyte-stimulating hormone receptor, melanotropin receptor, MSHR, MSH-RMGC14337, SHEP2 | |
| A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human Melanocortin-1 R/MC1R (NP_002377.4). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR, Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 4157 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title