missing translation for 'onlineSavingsMsg'
Learn More

Melanocortin-1 R/MC1R Rabbit anti-Human, Mouse, Rat, Clone: 6D1S5, Novus Biologicals™

Rabbit Monoclonal Antibody

£150.00 - £386.00

Specifications

Antigen Melanocortin-1 R/MC1R
Clone 6D1S5
Dilution Western Blot 1:500 - 1:2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18070844
View Documents
Novus Biologicals
NBP3-15409-20UL
20 μg
£150.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18328402
View Documents
Bio-Techne
NBP3-15409-100UL
100 μg
£386.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Melanocortin-1 R/MC1R Monoclonal antibody specifically detects Melanocortin-1 R/MC1R in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Melanocortin-1 R/MC1R
Western Blot 1:500 - 1:2000
Monoclonal
Purified
RUO
Human, Mouse, Rat
CMM5, MC1-R, melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor), Melanocortin receptor 1, melanocyte-stimulating hormone receptor, melanotropin receptor, MSHR, MSH-RMGC14337, SHEP2
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Melanocortin-1 R/MC1R (Q01726). MAVQGSQRRLLGSLNSTPTAIPQLGLAANQTGARCLEVSISDGLFLSLGLVSLVENALVVATIAKNRNLHSPMYCFICCLALSDLLVSGSNVLETAVILL
Primary
Store at -20°C. Avoid freeze-thaw cycles.
6D1S5
Western Blot
Unconjugated
Rabbit
GPCR, Neuroscience
PBS, 0.05% BSA, 50% glycerol, pH7.3
4157
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.