missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Melan-A Recombinant Rabbit Monoclonal Antibody (6V8D6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558028
This item is not returnable.
View return policy
Description
Immunogen sequence: VLLLIGCWYC RRRNGYRALM DKSLHVGTQC ALTRRCPQEG FDHRDSKVSL QEKNCEPVVP NAPPAYEKLS AEQSPPPYSP.
MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
Specifications
| Melan-A | |
| Recombinant Monoclonal | |
| 1.78 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q16655 | |
| MLANA | |
| A synthetic peptide corresponding to a sequence within amino acids 11-110 of human Melan-A/MART-1 (NP_005502.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 6V8D6 | |
| Unconjugated | |
| MLANA | |
| A930034P04Rik; Antigen LB39-AA; antigen SK29-AA; MART1; MART-1; melan-A; melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; MLANA; Protein Melan-A | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2315 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction