missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Melan-A Recombinant Rabbit Monoclonal Antibody (6V8D6)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558027

 View more versions of this product

Product Code. 30282692

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VLLLIGCWYC RRRNGYRALM DKSLHVGTQC ALTRRCPQEG FDHRDSKVSL QEKNCEPVVP NAPPAYEKLS AEQSPPPYSP.

MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Melan-A
Recombinant Monoclonal
1.78 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q16655
MLANA
A synthetic peptide corresponding to a sequence within amino acids 11-110 of human Melan-A/MART-1 (NP_005502.1).
20 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
6V8D6
Unconjugated
MLANA
A930034P04Rik; Antigen LB39-AA; antigen SK29-AA; MART1; MART-1; melan-A; melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; MLANA; Protein Melan-A
Rabbit
Protein A
RUO
2315
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.