Learn More
Invitrogen™ Melan-A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120982
Description
Positive test controls include: M21, Mouse eye, Rat eye. The target is usually found in the following locations: Endoplasmic reticulum membrane, Golgi apparatus, Melanosome, Single-pass type III membrane protein, trans-Golgi network membrane. Immunogen sequence: CRRRNGYRAL MDKSLHVGTQ CALTRRCPQE GFDHRDSKVS LQEKNCEPVV PNAPPAYEKL SAEQSPPPYS P.
MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
Specifications
| Melan-A | |
| Polyclonal | |
| Unconjugated | |
| MLANA | |
| A930034P04Rik; Antigen LB39-AA; antigen SK29-AA; MART1; MART-1; melan-A; melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; MLANA; Protein Melan-A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2315, 293890, 77836 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q16655 | |
| MLANA | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.