missing translation for 'onlineSavingsMsg'
Learn More

Melan-A/MART-1 Antibody (DT101 + BC199), Novus Biologicals™

Product Code. 18421622 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
Unit Size:
0.02mg
0.10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421622 0.1 mg 0.10mg
18440722 0.02 mg 0.02mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18421622 Supplier Novus Biologicals Supplier No. NBP2342460.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Melan-A/MART-1 Monoclonal antibody specifically detects Melan-A/MART-1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Melan-A/MART-1
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone DT101 + BC199
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. Q16655, Q16655, Q16655
Gene Alias Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART1MART-1, melan-A, melanoma antigen recognized by T-cells 1, Protein Melan-A
Host Species Mouse
Immunogen Recombinant hMelan-A/MART-1 protein (DT101 & BC199) (Uniprot: Q16655)
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2315
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG2b λ, IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.