missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ Melamine Compression Moulded Cap

Product Code. 10587862
Change view
Click to view available options
For Use With (Equipment):
250 to 1000 mL Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10587862 250 to 1000 mL Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10587862 Supplier Kautex™ Supplier No. 2000071160

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Black
Material Melamine
Thread Size (Metric) 00 mm
Neck Finish 00 mm
Product Type Compression Cap
For Use With (Equipment) 250 to 1000 mL Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.