missing translation for 'onlineSavingsMsg'
Learn More

Melamine, 99%

Product Code. p-3736800 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Unit Size:
100g
10kg
1kg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10765771 5 g Glass bottle 5g
10584584 100 g Plastic bottle 100g
10755771 1 kg Plastic bottle 1kg
10571964 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10765771 Supplier Thermo Scientific Acros Supplier No. 125350050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-78-1 | C3H6N6 | 126.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-78-1
Molecular Formula C3H6N6
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006055
InChI Key JDSHMPZPIAZGSV-UHFFFAOYSA-N
Synonym melamine, cyanurotriamide, cyanuramide, cyanurotriamine, isomelamine, theoharn, teoharn, triaminotriazine, cyanuric triamide, hicophor pr
PubChem CID 7955
ChEBI CHEBI:27915
IUPAC Name 1,3,5-triazine-2,4,6-triamine
SMILES NC1=NC(N)=NC(N)=N1

Specifications

Melting Point 354°C
Color White
Density 1.5730g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Beilstein 26, 245
Merck Index 15, 5880
Specific Gravity 1.573
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: 0.6g/L in ethanol (20°C),sparingly soluble in glycol,glycerol,pyridine,insoluble in ether,benzene,carbon tetrachloride
Formula Weight 126.12
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Melamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.