missing translation for 'onlineSavingsMsg'
Learn More

Melamine 1.0 mg/ml in Dimethyl Sulfoxide, LoGiCal

Product Code. 30460012
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30460012 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30460012

Brand: LoGiCal LGCAMP0056.0429

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Melamine 1.0 mg/ml in Dimethyl Sulfoxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Melamine
CAS 108-78-1
Analyte or Component Names Melamine
Shipping Condition Room Temperature
Molecular Formula C3 H6 N6
Certifications/Compliance ISO 17034
Synonym 1,3,5-Triazine-2,4,6-triamine, Metformin Hydrochloride Imp. D (EP), Melamine, Metformin Imp. D (EP), Metformin Hydrochloride Impurity D, Metformin Impurity D
InChI Formula InChI=1 S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)
SMILES Nc1nc(N)nc(N)n1
IUPAC Name 1,3,5-triazine-2,4,6-triamine
Solvent or Matrix Dimethyl Sulfoxide
Industry Type Forensics
Molecular Weight (g/mol) 126.12
Quantity 1 mL
Formula Weight 126.0654 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.