missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEKK6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104210
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MAP3K6.
Mitogen-activated protein kinase (MAPK) signaling cascades include MAPK or extracellular signal-regulated kinase (ERK), MAPK kinase (MKK or MEK), and MAPK kinase kinase (MAPKKK or MEKK). MAPKK kinase/MEKK phosphorylates and activates its downstream protein kinase, MAPK kinase/MEK, which in turn activates MAPK. The kinases of these signaling cascades are highly conserved, and homologs exist in yeast, Drosophila, and mammalian cells. MEKK6 activates the JNK, but not ERK or p38 kinase pathways.
Specifications
| MEKK6 | |
| Polyclonal | |
| Unconjugated | |
| MAP3K6 | |
| apoptosis signal-regulating kinase 2; ASK2; map3k6; MAPKKK6; MEKK6; MGC125653; MGC20114; mitogen activated protein kinase kinase kinase 6; mitogen-activated protein kinase kinase kinase 6; OTTHUMP00000005063 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 53608, 9064 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95382, Q9WTR2 | |
| MAP3K6 | |
| A synthesized peptide derived from human MAP3K6(Accession O95382), corresponding to amino acid residues A1252-P1288. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction