missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEKK4 Recombinant Rabbit Monoclonal Antibody (23GB4045), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | MEKK4 |
|---|---|
| Clone | 23GB4045 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089778
![]() |
Invitrogen™
MA552654 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088154
|
Invitrogen™
MA552655 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEKK4 Recombinant Monoclonal antibody specifically detects MEKK4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| MEKK4 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O08648, Q9Y6R4 | |
| 26407, 308106, 4216 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4045 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| D17Rp17; D17Rp17e; dJ473J16.1 (mitogen-activated protein kinase kinase kinase 4); JNKK1; KIAA0213; MAP three kinase 1; MAP/ERK kinase kinase 4; MAP2K4; Map3k4; MAPK/ERK kinase kinase 4; MAPKKK4; MEK kinase 4; Mek4b; MEKK 4; MEKK4; mitogen activated protein kinase kinase kinase 4; mitogen-activated protein kinase kinase kinase 4; mKIAA0213; MTK1; predicted protein of HQ0412; PRKMK4; PRO0412; RP17; Rp17a; SAPK/ERK kinase 1; SERK1; SSK2/SSK22 MAP kinase kinase kinase, yeast, homolog of; Tas; T-associated sex reversal | |
| A synthesized peptide derived from human MAP3K4 (170-200AA). | |
| Primary | |
| -20°C | |
| MAP3K4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title