missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEKK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104501
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MAP3K3.
MAP3K3 (MEKK3) directly regulates MAPK8 and extracellular signal-regulated protein kinase (ERK) pathways by phosphorylating multiple MAP2Ks. Northern blot analysis revealed a 4. 6-kb transcript that appears to be ubiquitously expressed. This protein directly regulates the stress-activated protein kinase (SAPK) and extracellular signal-regulated protein kinase (ERK) pathways by activating SEK and MEK1/2 respectively; it does not regulate the p38 pathway. In cotransfection assays, it enhanced transcription from a nuclear factor kappa-B (NFKB)-dependent reporter gene, consistent with a role in the SAPK pathway. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications
| MEKK3 | |
| Polyclonal | |
| Unconjugated | |
| Map3k3 | |
| AW548911; MAP/ERK kinase kinase 3; MAP3K3; MAPK/ERK kinase kinase 3; MAPKKK3; MEK kinase 3; MEKK 3; Mekk3; mitogen activated protein kinase kinase kinase 3; mitogen-activated protein kinase kinase kinase 3; mKIAA4031 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26406, 4215 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q61084, Q99759 | |
| Map3k3 | |
| A synthesized peptide derived from human MAP3K3(Accession Q99759), corresponding to amino acid residues V76-N126. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction