missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEKK2 Recombinant Rabbit Monoclonal Antibody (24GB260)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557458
This item is not returnable.
View return policy
Description
MEKK2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
MAP3K2 (MEKK2) is a serine/threonine kinase that activates kinases involved in the MAP kinase signaling pathway. This kinase preferentially activates other kinases involved in the MAP kinase signaling pathway. This kinase has been shown to directly phosphorylate and activate Ikappa B kinases, and thus plays a role in NF-kappa B signaling pathway. This kinase has also been found to bind and activate protein kinase C-related kinase 2, which suggests its involvement in a regulated signaling process.
Specifications
| MEKK2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Map3k2 | |
| 2.7.11.25; 9630061B06Rik; AI585793; MAP/ERK kinase kinase 2; MAP3K2; MAPK/ERK kinase kinase 2; MAPKKK2; MEK kinase 2; MEKK 2; MEKK2; MEKK2B; mitogen activated protein kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase 2; protein kinase MEKK2b | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10746, 171492, 26405 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB260 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q61083, Q9Y2U5 | |
| Map3k2 | |
| A synthesized peptide derived from human MEKK2 (aa 1-54). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction