missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEKK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106249
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MAP3K2.
MAP3K2 (MEKK2) is a serine/threonine kinase that activates kinases involved in the MAP kinase signaling pathway. This kinase preferentially activates other kinases involved in the MAP kinase signaling pathway. This kinase has been shown to directly phosphorylate and activate Ikappa B kinases, and thus plays a role in NF-kappa B signaling pathway. This kinase has also been found to bind and activate protein kinase C-related kinase 2, which suggests its involvement in a regulated signaling process.
Specifications
| MEKK2 | |
| Polyclonal | |
| Unconjugated | |
| Map3k2 | |
| 2.7.11.25; 9630061B06Rik; AI585793; MAP/ERK kinase kinase 2; MAP3K2; MAPK/ERK kinase kinase 2; MAPKKK2; MEK kinase 2; MEKK 2; MEKK2; MEKK2B; mitogen activated protein kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase 2; protein kinase MEKK2b | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10746, 171492, 26405 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q61083, Q9Y2U5 | |
| Map3k2 | |
| A synthesized peptide derived from human MAP3K2(Accession Q9Y2U5), corresponding to amino acid residues S493-D543. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction