Learn More
Invitrogen™ MEKK2 Monoclonal Antibody (4B4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515908
Description
MA5-15908 targets MAP3K2 in indirect ELISA, ICC, IF and WB applications and shows reactivity with Human samples. The MA5-15908 immunogen is purified recombinant fragment of human MAP3K2 expressed in E. Coli. . MA5-15908 detects MAP3K2 which has a predicted molecular weight of approximately 70kDa.
MAP3K2 (MEKK2) is a serine/threonine kinase that activates kinases involved in the MAP kinase signaling pathway. This kinase preferentially activates other kinases involved in the MAP kinase signaling pathway. This kinase has been shown to directly phosphorylate and activate Ikappa B kinases, and thus plays a role in NF-kappa B signaling pathway. This kinase has also been found to bind and activate protein kinase C-related kinase 2, which suggests its involvement in a regulated signaling process.
Specifications
| MEKK2 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| Q61083, Q9Y2U5 | |
| Map3k2 | |
| Purified recombinant fragment of human MAP3K2 expressed in E. Coli. | |
| RUO | |
| 10746, 26405 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 4B4 | |
| Unconjugated | |
| Map3k2 | |
| 2.7.11.25; 9630061B06Rik; AI585793; MAP/ERK kinase kinase 2; MAP3K2; MAPK/ERK kinase kinase 2; MAPKKK2; MEK kinase 2; MEKK 2; MEKK2; MEKK2B; mitogen activated protein kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase 2; protein kinase MEKK2b | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.